1-Methylcyclohexanecarbonyl chloride

Lieferant: Apollo Scientific

OR302386-1G OR302386-250MG OR302386-500MG
APOSOR302386-1GEA 0 EUR
APOSOR302386-1G APOSOR302386-250MG APOSOR302386-500MG
1-Methylcyclohexanecarbonyl chloride
1-Methylcyclohexanecarbonyl chloride
Formel: C₈H₁₃ClO
Lagertemperatur: Raumtemperatur
MDL: MFCD00019346
CAS-Nummer: 2890-61-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR