3-Amino-2-chloro-4-nitropyridine
Lieferant: Apollo Scientific
|
Achtung
|
OR302392-1G
APOSOR302392-1GEA
696
EUR
APOSOR302392-1G
3-Amino-2-chloro-4-nitropyridine
3-Amino-2-chloro-4-nitropyridine
Formel:
C₅H₄ClN₃O₂ Molecular Weight: 173,557 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD11656229 CAS-Nummer: 1092352-67-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...