4-Nitropyridin-3-amine

Lieferant: Apollo Scientific

OR302393-1G
APOSOR302393-1GEA 290 EUR
APOSOR302393-1G
4-Nitropyridin-3-amine
4-Nitropyridin-3-amine
Formel: C₅H₅N₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD04114232
CAS-Nummer: 13505-02-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR