4-[2-(Morpholin-4-yl)ethyl]aniline

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(2-Morpholinoethyl)aniline

OR302403-1G
APOSOR302403-1GEA 71.2 EUR
APOSOR302403-1G
4-[2-(Morpholin-4-yl)ethyl]aniline
4-[2-(Morpholin-4-yl)ethyl]aniline
Formel: C₁₂H₁₈N₂O
Molecular Weight: 206,284 g/mol
Schmelzpunkt: 92…94 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04114508
CAS-Nummer: 262368-47-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR