Methyl-6-methylpiperidine-3-carboxylate

Lieferant: Apollo Scientific

Synonyme: Methyl 6-methylpiperidine-3-carboxylate, 6-Methyl-3-piperidinecarboxylic acid methyl ester

OR302409-1G
APOSOR302409-1GEA 301 EUR
APOSOR302409-1G
Methyl-6-methylpiperidine-3-carboxylate
Methyl-6-methylpiperidine-3-carboxylate
Formel: C₈H₁₅NO₂
Molecular Weight: 157,21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13178893
CAS-Nummer: 908245-03-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR