3-(1H-1,2,4-Triazol-1-yl)piperidine 95%

Lieferant: Apollo Scientific

OR302416-1G OR302416-250MG
APOSOR302416-1GEA 944 EUR
APOSOR302416-1G APOSOR302416-250MG
3-(1H-1,2,4-Triazol-1-yl)piperidine 95%
3-(1H-1,2,4-Triazol-1-yl)piperidine
Formel: C₇H₁₂N₄
Molecular Weight: 152.197 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14632032
CAS-Nummer: 774511-83-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR