3-(1H-1,2,4-Triazol-1-yl)piperidine 95%
Lieferant: Apollo Scientific
OR302416-1G
OR302416-250MG
APOSOR302416-1GEA
944
EUR
APOSOR302416-1G
APOSOR302416-250MG
3-(1H-1,2,4-Triazol-1-yl)piperidine 95%
3-(1H-1,2,4-Triazol-1-yl)piperidine
Formel:
C₇H₁₂N₄ Molecular Weight: 152.197 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD14632032 CAS-Nummer: 774511-83-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...