4-Brompyrimidin-2-carbonitril

Lieferant: Apollo Scientific
Achtung

OR302429-1G
APOSOR302429-1GEA 928 EUR
APOSOR302429-1G
4-Brompyrimidin-2-carbonitril
4-Brompyrimidin-2-carbonitril
Formel: C₅H₂BrN₃
Lagertemperatur: Raumtemperatur
MDL: MFCD12400798
CAS-Nummer: 1160995-48-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR