4-[(Morpholin-4-yl)methyl]benzene-1,2-diamine

Lieferant: Apollo Scientific
Achtung

OR302434-1G
APOSOR302434-1GEA 603 EUR
APOSOR302434-1G
4-[(Morpholin-4-yl)methyl]benzene-1,2-diamine
4-[(Morpholin-4-yl)methyl]benzene-1,2-diamine
Formel: C₁₁H₁₇N₃O
Molecular Weight: 207,272 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11046432
CAS-Nummer: 825619-02-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR