Dihydrothiophen-3(2H)-one 1,1-dioxide

Lieferant: Apollo Scientific

OR302453-1G
APOSOR302453-1GEA 227 EUR
APOSOR302453-1G
Dihydrothiophen-3(2H)-one 1,1-dioxide
Dihydrothiophen-3(2H)-one 1,1-dioxide
Formel: C₄H₆O₃S
Molecular Weight: 134,154 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00154869
CAS-Nummer: 17115-51-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR