1-Phenylcyclopropanamine 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Phenylcyclopropylamine

OR302455-1G OR302455-5G
APOSOR302455-1GEA 59.6 EUR
APOSOR302455-1G APOSOR302455-5G
1-Phenylcyclopropanamine 95%
1-Phenylcyclopropanamine
Formel: C₉H₁₁N
Molecular Weight: 133.19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05863520
CAS-Nummer: 41049-53-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR