tert-Butyl (6-(hydroxymethyl)pyrimidin-4-yl)carbamate
Lieferant: Apollo Scientific
OR302465-1G
APOSOR302465-1GEA
935
EUR
APOSOR302465-1G
tert-Butyl (6-(hydroxymethyl)pyrimidin-4-yl)carbamate
tert-Butyl (6-(hydroxymethyl)pyrimidin-4-yl)carbamate
Formel:
C₁₀H₁₅N₃O₃ Molecular Weight: 225,244 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09952358 CAS-Nummer: 954097-20-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...