3,3-Dimethyl-5-nitroindolin-2-one

Lieferant: Apollo Scientific

OR302466-1G
APOSOR302466-1GEA 388 EUR
APOSOR302466-1G
3,3-Dimethyl-5-nitroindolin-2-one
3,3-Dimethyl-5-nitroindolin-2-one
Formel: C₁₀H₁₀N₂O₃
Molecular Weight: 206,198 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13191795
CAS-Nummer: 100511-00-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR