4-(2-Chlorothieno[3,2-d]pyrimidin-4-yl)morpholine

Lieferant: Apollo Scientific

OR302485-1G
APOSOR302485-1GEA 120 EUR
APOSOR302485-1G
4-(2-Chlorothieno[3,2-d]pyrimidin-4-yl)morpholine
4-(2-Chlorothieno[3,2-d]pyrimidin-4-yl)morpholine
Formel: C₁₀H₁₀ClN₃OS
Molecular Weight: 255,7200 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09909656
CAS-Nummer: 16234-15-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR