5-Bromoquinolin-2(1H)-one

Lieferant: Apollo Scientific

OR302486-1G
APOSOR302486-1GEA 462 EUR
APOSOR302486-1G
5-Bromoquinolin-2(1H)-one
5-Bromoquinolin-2(1H)-one
Formel: C₉H₆BrNO
Molecular Weight: 224,054 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11108678
CAS-Nummer: 99465-09-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR