5-(tert-Butyl)pyrazine-2-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR302488-1G
APOSOR302488-1GEA 945 EUR
APOSOR302488-1G
5-(tert-Butyl)pyrazine-2-carboxylic acid
5-(tert-Butyl)pyrazine-2-carboxylic acid
Formel: C₉H₁₂N₂O₂
Molecular Weight: 180,204 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20639242
CAS-Nummer: 242813-48-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR