6-Methoxy-1H-indazol

Lieferant: Apollo Scientific

OR302496-1G
APOSOR302496-1GEA 105 EUR
APOSOR302496-1G
6-Methoxy-1H-indazol
6-Methoxy-1H-indazole
Formel: C₈H₈N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD09261132
CAS-Nummer: 3522-07-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR