6-Morpholinopyrimidin-4-amine
Lieferant: Apollo Scientific
Synonyme:
6-Morpholinopyrimidin-4-amine, 4-Amino-6-morpholinopyrimidine
OR302503-1G
APOSOR302503-1GEA
120
EUR
APOSOR302503-1G
6-Morpholinopyrimidin-4-amine
6-Morpholinopyrimidin-4-amine
Formel:
C₈H₁₂N₄O Molecular Weight: 180,2071 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09999228 CAS-Nummer: 96225-80-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...