Pyridazine-3,6-diamine
Lieferant: Apollo Scientific
OR302511-1G
APOSOR302511-1GEA
98.6
EUR
APOSOR302511-1G
Pyridazine-3,6-diamine
Pyridazine-3,6-diamine
Formel:
C₄H₆N₄ Molecular Weight: 110,117 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD05665950 CAS-Nummer: 61070-99-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...