6-Morpholinopyrazin-2-amine

Lieferant: Apollo Scientific

OR302514-1G
APOSOR302514-1GEA 106 EUR
APOSOR302514-1G
6-Morpholinopyrazin-2-amine
6-Morpholinopyrazin-2-amine
Formel: C₈H₁₂N₄O
Lagertemperatur: Raumtemperatur
MDL: MFCD05722015
CAS-Nummer: 717847-03-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR