6-Methyl-2,4-pyrimidinediamine

Lieferant: Apollo Scientific

OR302527-1G
APOSOR302527-1GEA 102 EUR
APOSOR302527-1G
6-Methyl-2,4-pyrimidinediamine
6-Methyl-2,4-pyrimidinediamine
Formel: C₅H₈N₄
Molecular Weight: 124,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00127861
CAS-Nummer: 1791-73-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR