(4-Aminopyridin-3-yl)methanol
Lieferant: Apollo Scientific
Synonyme:
4-Amino-3-pyridinemethanol, (4-Amino-pyridin-3-yl)-methanol
OR302533-1G
APOSOR302533-1GEA
95.7
EUR
APOSOR302533-1G
(4-Aminopyridin-3-yl)methanol
(4-Aminopyridin-3-yl)methanol
Formel:
C₆H₈N₂O Molecular Weight: 124,14 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06203066 CAS-Nummer: 138116-34-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...