2-Isopropyl-6-methylpyrimidin-4-amine

Lieferant: Apollo Scientific

OR302540-1G
APOSOR302540-1GEA 289 EUR
APOSOR302540-1G
2-Isopropyl-6-methylpyrimidin-4-amine
2-Isopropyl-6-methylpyrimidin-4-amine
Formel: C₈H₁₃N₃
Molecular Weight: 151,209 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12189389
CAS-Nummer: 4595-70-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR