4-Amino-2-nitropyridine

Lieferant: Apollo Scientific

Synonyme: 2-Nitropyridin-4-amine

OR302545-1G
APOSOR302545-1GEA 302 EUR
APOSOR302545-1G
4-Amino-2-nitropyridine
4-Amino-2-nitropyridine
Formel: C₅H₅N₃O₂
Molecular Weight: 139,11 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04114153
CAS-Nummer: 14916-64-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR