6-Amino-3,4-dihydroisoquinolin-1(2H)-one

Lieferant: Apollo Scientific
Achtung

OR302549-1G
APOSOR302549-1GEA 719 EUR
APOSOR302549-1G
6-Amino-3,4-dihydroisoquinolin-1(2H)-one
6-Amino-3,4-dihydroisoquinolin-1(2H)-one
Formel: C₉H₁₀N₂O
Molecular Weight: 162,189 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11007917
CAS-Nummer: 22246-00-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR