6-(4-Methylpiperazin-1-yl)pyridazin-3-amine

Lieferant: Apollo Scientific

OR302562-1G
APOSOR302562-1GEA 306 EUR
APOSOR302562-1G
6-(4-Methylpiperazin-1-yl)pyridazin-3-amine
6-(4-Methylpiperazin-1-yl)pyridazin-3-amine
Formel: C₉H₁₅N₅
Molecular Weight: 193,249 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13192341
CAS-Nummer: 66346-94-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR