4-tert-Butylphenylisothiocyanat

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-tert-Butylphenylisothiocyanate

OR30261-1G
APOSOR30261-1GEA 21.9 EUR
APOSOR30261-1G
4-tert-Butylphenylisothiocyanat
4-tert-Butylphenylisothiocyanat
Formel: C₁₁H₁₃NS
Molecular Weight: 191,2926 g/mol
Siedepunkt: 92…94 °C/0,5mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD00022059
CAS-Nummer: 19241-24-8
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR