4-Bromo-5-methylpicolinaldehyde 97%

Lieferant: Apollo Scientific

OR302618-250MG
APOSOR302618-250MGEA 183 EUR
APOSOR302618-250MG
4-Bromo-5-methylpicolinaldehyde 97%
4-Bromo-5-methylpicolinaldehyde
Formel: C₇H₆BrNO
Molecular Weight: 200,033 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13190304
CAS-Nummer: 1196157-14-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR