4-Chloro-5-methylbenzene-1,2-diamine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Chloro-4,5-diamino-2-methylbenzene, 2-Chloro-4,5-diaminotoluene
OR30268-1G
APOSOR30268-1GEA
29
EUR
APOSOR30268-1G
4-Chloro-5-methylbenzene-1,2-diamine
4-Chloro-5-methylbenzene-1,2-diamine
Formel:
C₇H₉ClN₂ Molecular Weight: 156,613 g/mol Schmelzpunkt: 138…140 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00221471 CAS-Nummer: 63155-04-4 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...