(9aS)-Octahydropiperazino[2,1-c]morpholine dihydrochloride 97%

Lieferant: Apollo Scientific

Synonyme: (S)-Octahydropyrazino[2,1-c][1,4]oxazine dihydrochloride

OR302681-1G
APOSOR302681-1GEA 560 EUR
APOSOR302681-1G
(9aS)-Octahydropiperazino[2,1-c]morpholine dihydrochloride 97%
(9aS)-Octahydropiperazino[2,1-c]morpholine dihydrochloride
Formel: C₇H₁₄N₂O·2HCl
Molecular Weight: 215,121 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23105798
CAS-Nummer: 1089280-14-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR