(3aR,6aR)-tert-Butyl hexahydropyrrolo[3,4-b]pyrrole-5(1H)-carboxylate

Lieferant: Apollo Scientific
Achtung

OR302700-250MG
APOSOR302700-250MGEA 165 EUR
APOSOR302700-250MG
(3aR,6aR)-tert-Butyl hexahydropyrrolo[3,4-b]pyrrole-5(1H)-carboxylate
(3aR,6aR)-tert-Butyl hexahydropyrrolo[3,4-b]pyrrole-5(1H)-carboxylate
Formel: C₁₁H₂₀N₂O₂
Molecular Weight: 212,289 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18374593
CAS-Nummer: 370882-39-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR