(R)-2-(1-(tert-Butoxycarbonyl)pyrrolidin-2-yl)acetic acid

Lieferant: Apollo Scientific

OR302705-5G
APOSOR302705-5GEA 195 EUR
APOSOR302705-5G
(R)-2-(1-(tert-Butoxycarbonyl)pyrrolidin-2-yl)acetic acid
(R)-2-(1-(tert-Butoxycarbonyl)pyrrolidin-2-yl)acetic acid
Formel: C₁₁H₁₉NO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD06202402
CAS-Nummer: 101555-60-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR