2-Amino-4-methylbenzamid

Lieferant: Apollo Scientific

OR30273-1G
APOSOR30273-1GEA 65.4 EUR
APOSOR30273-1G
2-Amino-4-methylbenzamid
2-Amino-4-methylbenzamid
Formel: C₈H₁₀N₂O
Molecular Weight: 150,1800 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00221474
CAS-Nummer: 39549-79-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR