(1S,5R)-3-tert-Butoxycarbonyl-3-azabicyclo[3.1.0]hexane-6-carboxylic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: rel-(1R,5S,6r)-3-(tert-Butoxycarbonyl)-3-azabicyclo[3.1.0]hexane-6-carboxylic acid

OR302739-1G
APOSOR302739-1GEA 79.9 EUR
APOSOR302739-1G
(1S,5R)-3-tert-Butoxycarbonyl-3-azabicyclo[3.1.0]hexane-6-carboxylic acid
(1S,5R)-3-tert-Butoxycarbonyl-3-azabicyclo[3.1.0]hexane-6-carboxylic acid
Formel: C₁₁H₁₇NO₄
Molecular Weight: 227,26 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12198680
CAS-Nummer: 927679-54-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR