Oxetan-3-yl methanesulphonate

Lieferant: Apollo Scientific

OR302751-1G
APOSOR302751-1GEA 198 EUR
APOSOR302751-1G
Oxetan-3-yl methanesulphonate
Oxetan-3-yl methanesulphonate
Formel: C₄H₈O₄S
Lagertemperatur: Raumtemperatur
MDL: MFCD22192544
CAS-Nummer: 148430-81-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR