1-Propyl-1H-1,2,3-triazol-4-amine

Lieferant: Apollo Scientific

OR302763-1G
APOSOR302763-1GEA 168 EUR
APOSOR302763-1G
1-Propyl-1H-1,2,3-triazol-4-amine
1-Propyl-1H-1,2,3-triazol-4-amine
Formel: C₅H₁₀N₄
Molecular Weight: 126,16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08059914
CAS-Nummer: 915924-69-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR