2(1H-Benzoimidazol-2-yl)-pyridin-4-yl amine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(1H-Benzo[d]imidazol-2-yl)pyridin-4-amine
OR302765-250MG
APOSOR302765-250MGEA
219
EUR
APOSOR302765-250MG
2(1H-Benzoimidazol-2-yl)-pyridin-4-yl amine
2(1H-Benzoimidazol-2-yl)-pyridin-4-yl amine
Formel:
C₁₂H₁₀N₄ Molecular Weight: 210,235 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11107322 CAS-Nummer: 1082192-60-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...