4-Spiro[1-phthalan]piperidine hydrochloride

Lieferant: Apollo Scientific

Synonyme: 3H-Spiro[isobenzofuran-1,4'-piperidine] hydrochloride

OR302767-1G
APOSOR302767-1GEA 94.4 EUR
APOSOR302767-1G
4-Spiro[1-phthalan]piperidine hydrochloride
4-Spiro[1-phthalan]piperidine hydrochloride
Formel: C₁₂H₁₆ClNO
Molecular Weight: 225,72 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02179146
CAS-Nummer: 37663-44-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR