N-[2-(Acetylamino)phenyl]acetamide
Lieferant: Apollo Scientific
Synonyme:
N,N'-(1,2-Phenylene)diacetamide
OR30279-1G
APOSOR30279-1GEA
89.9
EUR
APOSOR30279-1G
N-[2-(Acetylamino)phenyl]acetamide
N-[2-(Acetylamino)phenyl]acetamide
Formel:
C₁₀H₁₂N₂O₂ Molecular Weight: 192,22 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00092752 CAS-Nummer: 2050-85-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...