1-[(4-Isothiocyanatophenyl)sulphonyl]piperidine

Lieferant: Apollo Scientific

OR30283-1G
APOSOR30283-1GEA 168 EUR
APOSOR30283-1G
1-[(4-Isothiocyanatophenyl)sulphonyl]piperidine
1-[(4-Isothiocyanatophenyl)sulphonyl]piperidine
Formel: C₁₂H₁₄N₂O₂S₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00023666
CAS-Nummer: 7356-55-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR