5-Butyl-2-iodtoluol

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Butyl-1-iod-2-methylbenzol

OR30290-10G
APOSOR30290-10GEA 0 EUR
APOSOR30290-10G
5-Butyl-2-iodtoluol
5-Butyl-2-iodtoluol
Formel: C₁₁H₁₅I
Molecular Weight: 274.14 g/mol
Siedepunkt: 80…82 °C/0.1mmHg
Lagertemperatur: Raumtemperatur
CAS-Nummer: 175278-38-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR