2-Chlor-4-nitrobenzamid

Lieferant: Apollo Scientific
Achtung

OR30292-1G
APOSOR30292-1GEA 26.1 EUR
APOSOR30292-1G
2-Chlor-4-nitrobenzamid
2-Chlor-4-nitrobenzamid
Formel: C₇H₅ClN₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00017119
CAS-Nummer: 3011-89-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR