3-(4-tert-Butylphenyl)-1-methyl-1H-pyrazol-5-amine

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Amino-3-[4-(tert-butyl)phenyl]-1-methyl-1H-pyrazole, 1-(5-Amino-1-methyl-1H-pyrazol-3-yl)-4-(tert-butyl)benzene

OR3030-250MG
APOSOR3030-250MGEA 122 EUR
APOSOR3030-250MG
3-(4-tert-Butylphenyl)-1-methyl-1H-pyrazol-5-amine
3-(4-tert-Butylphenyl)-1-methyl-1H-pyrazol-5-amine
Formel: C₁₄H₁₉N₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00067993
CAS-Nummer: 175137-47-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR