1-Carbamoylcyclopropane-1-carboxylic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Carbamoylcyclopropanecarboxylic acid

OR303011-1G
APOSOR303011-1GEA 26.1 EUR
APOSOR303011-1G
1-Carbamoylcyclopropane-1-carboxylic acid
1-Carbamoylcyclopropane-1-carboxylic acid
Formel: C₅H₇NO₃
Molecular Weight: 129.12 g/mol
Schmelzpunkt: 185 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00190650
CAS-Nummer: 6914-74-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR