2-Chloro-1-methyl-5-nitro-1H-1,3-benzodiazole
Lieferant: Apollo Scientific
OR303012-500MG
APOSOR303012-500MGEA
438
EUR
APOSOR303012-500MG
2-Chloro-1-methyl-5-nitro-1H-1,3-benzodiazole
2-Chloro-1-methyl-5-nitro-1H-1,3-benzodiazole
Formel:
C₈H₆ClN₃O₂ Molecular Weight: 211.61 g/mol Schmelzpunkt: 180…182 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09953108 CAS-Nummer: 15965-66-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...