(2,5-Dimethyl-1,3-thiazol-4-yl)acetic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(2,5-Dimethylthiazol-4-yl)acetic acid

OR30302-1G
APOSOR30302-1GEA 75.4 EUR
APOSOR30302-1G
(2,5-Dimethyl-1,3-thiazol-4-yl)acetic acid
(2,5-Dimethyl-1,3-thiazol-4-yl)acetic acid
Formel: C₇H₉NO₂S
Molecular Weight: 171,22 g/mol
Schmelzpunkt: 110…113 °C
Lagertemperatur: Kühlschrank
MDL: MFCD02677725
CAS-Nummer: 306937-38-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR