6-[(tert-Butyldimethylsilyl)oxy]-1H-indole
Lieferant: Apollo Scientific
OR303045-500MG
APOSOR303045-500MGEA
336
EUR
APOSOR303045-500MG
6-[(tert-Butyldimethylsilyl)oxy]-1H-indole
6-[(tert-Butyldimethylsilyl)oxy]-1H-indole
Formel:
C₁₄H₂₁NOSi Lagertemperatur: Raumtemperatur |
MDL:
MFCD02683922 CAS-Nummer: 106792-41-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...