Benzyl-4-[methoxy(methyl)carbamoyl]piperidine-1-carboxylate
Lieferant: Apollo Scientific
OR303056-1G
APOSOR303056-1GEA
128
EUR
APOSOR303056-1G
Benzyl-4-[methoxy(methyl)carbamoyl]piperidine-1-carboxylate
Benzyl-4-[methoxy(methyl)carbamoyl]piperidine-1-carboxylate
Formel:
C₁₆H₂₂N₂O₄ Molecular Weight: 306.36 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07370044 CAS-Nummer: 148148-48-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...