2-Chlorobenzene-1-carboximidamide
Lieferant: Apollo Scientific
|
Achtung
|
OR303066-1G
APOSOR303066-1GEA
638
EUR
APOSOR303066-1G
2-Chlorobenzene-1-carboximidamide
2-Chlorobenzene-1-carboximidamide
Formel:
C₇H₇ClN₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD05662890 CAS-Nummer: 45743-05-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...