(6-Chloro-4-methoxypyridin-3-yl)methanol 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Chloro-5-(hydroxymethyl)-4-methoxypyridine
OR303070-250MG
APOSOR303070-250MGEA
79.9
EUR
APOSOR303070-250MG
(6-Chloro-4-methoxypyridin-3-yl)methanol 97%
(6-Chloro-4-methoxypyridin-3-yl)methanol
Formel:
C₇H₈ClNO₂ Molecular Weight: 173,6 g/mol Schmelzpunkt: 107…108 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD22551808 CAS-Nummer: 1263059-66-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...